accessibility menu, dialog, popup

UserName

CAS RN 34289-60-6

IUPAC Name: hydrogen 4,6-dimethyl-1,2-dihydropyrimidin-2-one chloride
Synonyms: hydrogen 4,6-dimethyl-1H-pyrimidin-2-one chloride
Molecular Weight (g/mol): 160.6
Molecular Formula: C6H9ClN2O
InChi Key: IFXXETYYSLJRNF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC(C)=NC(=O)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results