missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethyl-2-hydroxypyrimidine Hydrochloride 98.0+%, TCI America™

Catalog No. D182325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D182325G 25 g
D1823250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D182325G Supplier TCI America Supplier No. D182325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34289-60-6
Molecular Formula C6H9ClN2O
Molecular Weight (g/mol) 160.601
MDL Number MFCD00035312
InChI Key IFXXETYYSLJRNF-UHFFFAOYSA-N
Synonym 4,6-Dimethyl-2-pyrimidinol Hydrochloride, 2-Hydroxy-4,6-dimethylpyrimidine Hydrochloride
PubChem CID 12343166
IUPAC Name 4,6-dimethyl-1H-pyrimidin-2-one;hydrochloride
SMILES CC1=CC(=NC(=O)N1)C.Cl

Specifications

Melting Point 281°C
Quantity 25 g
Formula Weight 160.60
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dimethyl-2-hydroxypyrimidine Hydrochloride
EINECSNumber (9)-2245
TSCA No