missing translation for 'onlineSavingsMsg'
Learn More

4',6'-Dimethoxy-2'-hydroxyacetophenone 98.0+%, TCI America™

Catalog No. p-7127439
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26831G 1 g
D26835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26831G Supplier TCI America Supplier No. D26831G
Only null left

Chemical Identifiers

CAS 90-24-4
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00017243
InChI Key FBUBVLUPUDBFME-UHFFFAOYSA-N
Synonym xanthoxylin, 2'-hydroxy-4',6'-dimethoxyacetophenone, brevifolin, xanthoxyline, 2-hydroxy-4,6-dimethoxyacetophenone, 1-2-hydroxy-4,6-dimethoxyphenyl ethanone, brevifolin zanthoxylum, phloracetophenone dimethyl ether, brevifolin van, 1-2-hydroxy-4,6-dimethoxyphenyl ethan-1-one
PubChem CID 66654
ChEBI CHEBI:10070
IUPAC Name 1-(2-hydroxy-4,6-dimethoxyphenyl)ethan-1-one
SMILES COC1=CC(O)=C(C(C)=O)C(OC)=C1

Specifications

Melting Point 81°C
Boiling Point 185°C
Quantity 1 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′,6′-Dimethoxy-2′-hydroxyacetophenone