missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Dihydroxypyrimidine 95.0+%, TCI America™
Supplier: TCI America D1821250G
Specifications
| 4,6-Dihydroxypyrimidine | |
| C4H4N2O2 | |
| 250 g | |
| DUFGYCAXVIUXIP-UHFFFAOYSA-N | |
| 6-hydroxy-3,4-dihydropyrimidin-4-one | |
| 14512 | |
| ≥95.0% (T) |
| 1193-24-4 | |
| MFCD00016733 | |
| 4,6-dihydroxypyrimidine, pyrimidine-4,6-diol, 4,6-pyrimidinediol, 6-hydroxy-4 1h-pyrimidinone, 4 1h-pyrimidinone, 6-hydroxy, 4 3h-pyrimidinone, 6-hydroxy, 6-hydroxypyrimidin-4 3h-one, 6-hydroxy-1h-pyrimidin-4-one, 4,6-dihydroxypyrimidin, 6-hydroxy-4-pyrimidinone | |
| OC1=CC(=O)NC=N1 | |
| 112.09 | |
| 112.09 | |
| Crystalline Powder |
Chemical Identifiers
| 1193-24-4 | |
| 112.09 | |
| DUFGYCAXVIUXIP-UHFFFAOYSA-N | |
| 14512 | |
| OC1=CC(=O)NC=N1 |
| C4H4N2O2 | |
| MFCD00016733 | |
| 4,6-dihydroxypyrimidine, pyrimidine-4,6-diol, 4,6-pyrimidinediol, 6-hydroxy-4 1h-pyrimidinone, 4 1h-pyrimidinone, 6-hydroxy, 4 3h-pyrimidinone, 6-hydroxy, 6-hydroxypyrimidin-4 3h-one, 6-hydroxy-1h-pyrimidin-4-one, 4,6-dihydroxypyrimidin, 6-hydroxy-4-pyrimidinone | |
| 6-hydroxy-3,4-dihydropyrimidin-4-one |
Safety and Handling
RTECSNumber : UW7523000
TSCA : Yes