missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxypyrimidine 95.0+%, TCI America™

Catalog No. p-7126949
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D182125G 25 g
D1821250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D182125G Supplier TCI America Supplier No. D182125G
Only null left

Chemical Identifiers

CAS 1193-24-4
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00016733
InChI Key DUFGYCAXVIUXIP-UHFFFAOYSA-N
Synonym 4,6-dihydroxypyrimidine, pyrimidine-4,6-diol, 4,6-pyrimidinediol, 6-hydroxy-4 1h-pyrimidinone, 4 1h-pyrimidinone, 6-hydroxy, 4 3h-pyrimidinone, 6-hydroxy, 6-hydroxypyrimidin-4 3h-one, 6-hydroxy-1h-pyrimidin-4-one, 4,6-dihydroxypyrimidin, 6-hydroxy-4-pyrimidinone
PubChem CID 14512
IUPAC Name 6-hydroxy-3,4-dihydropyrimidin-4-one
SMILES OC1=CC(=O)NC=N1

Specifications

Quantity 25 g
Formula Weight 112.09
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dihydroxypyrimidine
RTECSNumber UW7523000
TSCA Yes