missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxy-5-nitropyrimidine 98.0+%, TCI America™

Catalog No. D196025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D196025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D196025G Supplier TCI America Supplier No. D196025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2164-83-2
Molecular Formula C4H3N3O4
Molecular Weight (g/mol) 157.09
MDL Number MFCD00006110
InChI Key ABTLZAVJDRUDNG-UHFFFAOYSA-N
Synonym 4,6-dihydroxy-5-nitropyrimidine, 5-nitropyrimidine-4,6-diol, 4,6-dihydroxy-5-nitropyrimdine, 5-nitro-4,6-pyrimidinediol, 4 1h-pyrimidinone, 6-hydroxy-5-nitro, 6-hydroxy-5-nitro-3h-pyrimidin-4-one, 6-hydroxy-5-nitro-4 1h-pyrimidinone, 5-nitro-4,6-dihydroxypyrimidine, 4-hydroxy-5-nitro-1h-pyrimidin-6-one, 4 3h-pyrimidinone, 6-hydroxy-5-nitro
PubChem CID 75105
IUPAC Name 6-hydroxy-5-nitro-3,4-dihydropyrimidin-4-one
SMILES OC1=C(C(=O)NC=N1)[N+]([O-])=O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 157.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dihydroxy-5-nitropyrimidine