accessibility menu, dialog, popup

UserName

OHONHNNOO

CAS RN 2164-83-2

IUPAC Name: 6-hydroxy-5-nitro-3,4-dihydropyrimidin-4-one
Synonyms: 6-hydroxy-5-nitro-3H-pyrimidin-4-one
Molecular Weight (g/mol): 157.09
Molecular Formula: C4H3N3O4
InChi Key: ABTLZAVJDRUDNG-UHFFFAOYSA-N
SMILES: OC1=C(C(=O)NC=N1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results