Learn More
4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98+%
CAS: 2465-59-0 | C5H4N4O2 | 152.113 g/mol
$80.68 - $506.64
Chemical Identifiers
| CAS | 2465-59-0 |
|---|---|
| Molecular Formula | C5H4N4O2 |
| Molecular Weight (g/mol) | 152.113 |
| MDL Number | MFCD00056934 |
| InChI Key | HXNFUBHNUDHIGC-UHFFFAOYSA-N |
| Synonym | oxypurinol, oxipurinol, alloxanthine, 1h-pyrazolo 3,4-d pyrimidine-4,6-diol, oxoallopurinol, 4,6-dihydroxypyrazolo 3,4-d pyrimidine, 1h-pyrazolo 3,4-d pyrimidine-4,6 5h,7h-dione, oxipurinolum, 1h-pyrazolo 3,4-d pyrimidine-4,6 2h,5h-dione, alloxanthin van |
| PubChem CID | 4644 |
| ChEBI | CHEBI:28315 |
| IUPAC Name | 1,2-dihydropyrazolo[3,4-d]pyrimidine-4,6-dione |
| SMILES | C1=C2C(=NC(=O)NC2=O)NN1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL07160MD
|
Thermo Scientific Chemicals
L07160MD |
250 mg |
Each for $80.68
|
|
|||||
|
AAL0716003
|
Thermo Scientific Chemicals
L0716003 |
1 g |
Each for $163.82
|
|
|||||
|
AAL0716006
|
Thermo Scientific Chemicals
L0716006 |
5 g |
Each for $506.64
|
|
|||||
Description
4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine is an inhibitor to study the specificity and kinetics of xanthine oxidase. It is also used as an anti-inflammatory agent via inhibition of xanthine oxidase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine is an inhibitor to study the specificity and kinetics of xanthine oxidase. It is also used as an anti-inflammatory agent via inhibition of xanthine oxidase.
Solubility
Soluble in DMSO.
Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
| 2465-59-0 | |
| 152.113 | |
| HXNFUBHNUDHIGC-UHFFFAOYSA-N | |
| 4644 | |
| 1,2-dihydropyrazolo[3,4-d]pyrimidine-4,6-dione |
| C5H4N4O2 | |
| MFCD00056934 | |
| oxypurinol, oxipurinol, alloxanthine, 1h-pyrazolo 3,4-d pyrimidine-4,6-diol, oxoallopurinol, 4,6-dihydroxypyrazolo 3,4-d pyrimidine, 1h-pyrazolo 3,4-d pyrimidine-4,6 5h,7h-dione, oxipurinolum, 1h-pyrazolo 3,4-d pyrimidine-4,6 2h,5h-dione, alloxanthin van | |
| CHEBI:28315 | |
| C1=C2C(=NC(=O)NC2=O)NN1 |
Specifications
| 2465-59-0 | |
| Odorless | |
| MFCD00056934 | |
| 139956 | |
| Soluble in DMSO. | |
| C1=C2C(=NC(=O)NC2=O)NN1 | |
| 152.113 | |
| CHEBI:28315 | |
| ≥98% |
| >300°C | |
| C5H4N4O2 | |
| 250 mg | |
| oxypurinol, oxipurinol, alloxanthine, 1h-pyrazolo 3,4-d pyrimidine-4,6-diol, oxoallopurinol, 4,6-dihydroxypyrazolo 3,4-d pyrimidine, 1h-pyrazolo 3,4-d pyrimidine-4,6 5h,7h-dione, oxipurinolum, 1h-pyrazolo 3,4-d pyrimidine-4,6 2h,5h-dione, alloxanthin van | |
| HXNFUBHNUDHIGC-UHFFFAOYSA-N | |
| 1,2-dihydropyrazolo[3,4-d]pyrimidine-4,6-dione | |
| 4644 | |
| 152.11 | |
| 4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 219-570-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only