accessibility menu, dialog, popup

UserName

CAS RN 2465-59-0

IUPAC Name: 1H,2H,4H,5H,6H-pyrazolo[3,4-d]pyrimidine-4,6-dione
Synonyms: oxypurinol 奥昔嘌醇
Molecular Weight (g/mol): 152.11
Molecular Formula: C5H4N4O2
InChi Key: HXNFUBHNUDHIGC-UHFFFAOYSA-N
SMILES: O=C1NC(=O)C2=CNNC2=N1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)