missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloropyrimidine-5-carboxaldehyde, 97%

Catalog No. p-7036194
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5444903 1 g
AAH5444906 5 g
AAH5444914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5444903 Supplier Thermo Scientific Chemicals Supplier No. H5444903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5305-40-8 | C5H2Cl2N2O | 176.98 g/mol

4,6-Dichloropyrimidine-5-carboxaldehyde is used as a substrate in the synthesis of N-terminal surrogate in amino acid and peptide analogues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dichloropyrimidine-5-carboxaldehyde is used as a substrate in the synthesis of N-terminal surrogate in amino acid and peptide analogues.

Notes
Incompatible with strong oxidizing agents and strong acids. Store in cool place.

Chemical Identifiers

CAS 5305-40-8
Molecular Formula C5H2Cl2N2O
Molecular Weight (g/mol) 176.98
MDL Number MFCD02257701
InChI Key XQSJHQXYQAUDFC-UHFFFAOYSA-N
Synonym 4,6-dichloro-5-pyrimidinecarbaldehyde, 4,6-dichloro-pyrimidine-5-carbaldehyde, 4,6-dichloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 4,6-dichloro, 4,6-dichloro-5-formylpyrimidine, 4,6-dichloro-5-pyrimidinecarboxaldehyde, 4,6-di chloro pyrimidine-5-aldehyde, 4,6-dichlorpyrimidin-5-carbaldehyd, zlchem 524, pubchem2117
PubChem CID 819691
IUPAC Name 4,6-dichloropyrimidine-5-carbaldehyde
SMILES ClC1=NC=NC(Cl)=C1C=O

Specifications

Melting Point 66°C to 71°C
Quantity 1 g
Formula Weight 175.99
Percent Purity 97%
Chemical Name or Material 4,6-Dichloropyrimidine-5-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only