accessibility menu, dialog, popup

UserName

CAS RN 5305-40-8

IUPAC Name: 4,6-dichloropyrimidine-5-carbaldehyde
Synonyms: 4,6-dichloropyrimidine-5-carbaldehyde
Molecular Weight (g/mol): 176.98
Molecular Formula: C5H2Cl2N2O
InChi Key: XQSJHQXYQAUDFC-UHFFFAOYSA-N
SMILES: ClC1=NC=NC(Cl)=C1C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results