missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloronicotinic acid, 97%

Catalog No. AAH6138106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6138106 5 g
AAH6138114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6138106 Supplier Thermo Scientific Chemicals Supplier No. H6138106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 73027-79-9 | C6H3Cl2NO2 | 191.995 g/mol

4,6-Dichloronicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dichloronicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Protect from heat. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 73027-79-9
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 191.995
MDL Number MFCD00234146
InChI Key ILMIEWNDXAKVNI-UHFFFAOYSA-N
Synonym 4,6-dichloronicotinic acid, 2,4-dichloro-5-carboxylpyridine, 4,6-dichloro nicotinic acid, 2,4-dichloropyridine-5-carboxylic acid, 3-pyridinecarboxylic acid, 4,6-dichloro, 2,4-dichloro-5-carboxypyridine, pubchem18521, acmc-209op3, 4.6-dichloronicotinic acid, 4,6-dichloro-nicotinic acid
PubChem CID 817158
IUPAC Name 4,6-dichloropyridine-3-carboxylic acid
SMILES C1=C(C(=CN=C1Cl)C(=O)O)Cl

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 192
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4,6-Dichloronicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only