accessibility menu, dialog, popup

UserName

CAS RN 73027-79-9

IUPAC Name: 4,6-dichloropyridine-3-carboxylic acid
Synonyms: 4,6-dichloropyridine-3-carboxylic acid
Molecular Weight (g/mol): 192
Molecular Formula: C6H3Cl2NO2
InChi Key: ILMIEWNDXAKVNI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results