missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-5-methylpyrimidine 98.0+%, TCI America™

Catalog No. D40915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40915G 5 g
D409125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40915G Supplier TCI America Supplier No. D40915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4316-97-6
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.00
MDL Number MFCD01646112
InChI Key NUEYDUKUIXVKNB-UHFFFAOYSA-N
Synonym 4,6-dichloro-5-methyl-pyrimidine, pyrimidine, 4,6-dichloro-5-methyl, pubchem13429, acmc-209jt4, ksc490s5p, 4,6-dichloro-5-methyl pyrimidine, 4,6-dichloro-5-methyl-1,3-diazine, 4,6-dichloro-5-methylpyrimidine
PubChem CID 818860
IUPAC Name 4,6-dichloro-5-methylpyrimidine
SMILES CC1=C(Cl)N=CN=C1Cl

Specifications

Melting Point 59°C
Quantity 5 g
Formula Weight 163.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dichloro-5-methylpyrimidine