accessibility menu, dialog, popup

UserName

CH3NNClCl

CAS RN 4316-97-6

IUPAC Name: 4,6-dichloro-5-methylpyrimidine
Synonyms: 4,6-dichloro-5-methylpyrimidine
Molecular Weight (g/mol): 163
Molecular Formula: C5H4Cl2N2
InChi Key: NUEYDUKUIXVKNB-UHFFFAOYSA-N
SMILES: CC1=C(Cl)N=CN=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results