missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-5-methoxypyrimidine 98.0+%, TCI America™

Catalog No. D47235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47235G 5 g
D472325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47235G Supplier TCI America Supplier No. D47235G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5018-38-2
Molecular Formula C5H4Cl2N2O
Molecular Weight (g/mol) 179
MDL Number MFCD00087680
InChI Key IJQIGKLDBGKSNT-UHFFFAOYSA-N
Synonym 4,6-dichlor-5-methoxypyrimidin, 4,6-dichloro-5-methoxy-pyrimidine, 4,6-dichloro-5-methoxy pyrimidine, zlchem 448, pubchem7077, pyrimidine, 4,6-dichloro-5-methoxy, acmc-1avm3, 4,6 dichloro-5-methoxypyrimidine, pyrimidine,6-dichloro-5-methoxy, 4,6-dichloro-5-methoxypyrimidine
PubChem CID 78720
IUPAC Name 4,6-dichloro-5-methoxypyrimidine
SMILES COC1=C(N=CN=C1Cl)Cl

Specifications

Melting Point 57°C
Color Yellow
Quantity 5 g
Formula Weight 179.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dichloro-5-methoxypyrimidine