accessibility menu, dialog, popup

UserName

H3COClNNCl

CAS RN 5018-38-2

IUPAC Name: 4,6-dichloro-5-methoxypyrimidine
Synonyms: 4,6-dichloro-5-methoxypyrimidine
Molecular Weight (g/mol): 179
Molecular Formula: C5H4Cl2N2O
InChi Key: IJQIGKLDBGKSNT-UHFFFAOYSA-N
SMILES: COC1=C(Cl)N=CN=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results