missing translation for 'onlineSavingsMsg'
Learn More

8-Aminoisoquinoline, 97%

Catalog No. AC429400050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429400050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC429400050 Supplier Thermo Scientific Chemicals Supplier No. 429400050
Only null left

CAS: 23687-27-6 | C9H8N2 | 144.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23687-27-6
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00179553
InChI Key GUSYANXQYUJOBH-UHFFFAOYSA-N
Synonym 8-aminoisoquinoline, 8-isoquinolinamine, isoquinolin-8-ylamine, 8-amino-isoquinoline, 8-isoquinolylamine, 8-amine-isoquinoline, pubchem6251, ksc201o9p, isoquinolin-8-amine; isoquinolin-8-ylamine
PubChem CID 13927912
IUPAC Name isoquinolin-8-amine
SMILES NC1=C2C=NC=CC2=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 168.0°C to 169.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 144.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 8-Aminoisoquinoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only