missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-5-fluoropyrimidine, 98%

Catalog No. p-7053967
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6177003 1 g
AAH6177006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6177003 Supplier Thermo Scientific Chemicals Supplier No. H6177003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 213265-83-9 | C4HCl2FN2 | 166.96 g/mol

4,6-Dichloro-5-Fluoropyrimidine is an useful intermediate for pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dichloro-5-Fluoropyrimidine is an useful intermediate for pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 213265-83-9
Molecular Formula C4HCl2FN2
Molecular Weight (g/mol) 166.96
MDL Number MFCD08056331
InChI Key DGMIGAHDDPJOPN-UHFFFAOYSA-N
Synonym 4,6-dichloro-5-fluoro-pyrimidine, pyrimidine, 4,6-dichloro-5-fluoro, 4,6-dichloro-5-fluoro-1,3-diazine, pubchem18606, acmc-209fj8, ksc494o9j, 4,6-dichloro-5-fluoropyrimidine;, 4,6-dichloro-5-fluoro-pyrimidine;, 5-fluoro-4,6-dichloropyrimidine, 4 3h-pyrimidinone,2-chloro-5-fluoro
PubChem CID 11286675
IUPAC Name 4,6-dichloro-5-fluoropyrimidine
SMILES FC1=C(Cl)N=CN=C1Cl

Specifications

Boiling Point 50°C to 60°C (14 mmHg)
Flash Point 91°C (196°F)
Quantity 1 g
UN Number UN3265
Solubility Information Slightly soluble in water.
Formula Weight 166.97
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4,6-Dichloro-5-fluoropyrimidine
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only