CAS RN 213265-83-9
IUPAC Name:
4,6-dichloro-5-fluoropyrimidine
Synonyms:
4,6-dichloro-5-fluoropyrimidine
Molecular Weight (g/mol):
166.96
Molecular Formula:
C4HCl2FN2
InChi Key:
DGMIGAHDDPJOPN-UHFFFAOYSA-N
SMILES:
FC1=C(Cl)N=CN=C1Cl
1
–
1
of
1
results
1
Promotions Available
4,6-Dichloro-5-fluoropyrimidine, 98%
4,6-Dichloro-5-fluoropyrimidine, 98%
CAS: 213265-83-9 Molecular Formula: C4HCl2FN2 Molecular Weight (g/mol): 166.96 MDL Number: MFCD08056331 InChI Key: DGMIGAHDDPJOPN-UHFFFAOYSA-N Synonym: 4,6-dichloro-5-fluoro-pyrimidine,pyrimidine, 4,6-dichloro-5-fluoro,4,6-dichloro-5-fluoro-1,3-diazine,pubchem18606,acmc-209fj8,ksc494o9j,4,6-dichloro-5-fluoropyrimidine;,4,6-dichloro-5-fluoro-pyrimidine;,5-fluoro-4,6-dichloropyrimidine,4 3h-pyrimidinone,2-chloro-5-fluoro PubChem CID: 11286675 IUPAC Name: 4,6-dichloro-5-fluoropyrimidine SMILES: FC1=C(Cl)N=CN=C1Cl