missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-2-(methylthio)pyrimidine 98.0+%, TCI America™

Catalog No. p-7127354
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25335G 5 g
D253325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25335G Supplier TCI America Supplier No. D25335G
Only null left

Chemical Identifiers

CAS 6299-25-8
Molecular Formula C5H4Cl2N2S
Molecular Weight (g/mol) 195.061
MDL Number MFCD00006086
InChI Key FCMLONIWOAGZJX-UHFFFAOYSA-N
Synonym 4,6-dichloro-2-methylthio pyrimidine, 4,6-dichloro-2-methylmercaptopyrimidine, 4,6-dichloro-2-methylsulfanyl-pyrimidine, 4,6-dichloro-2-methylthiopyrimidine, pyrimidine, 4,6-dichloro-2-methylthio, 4,6-dichloro-2-methylsulfanyl pyrimidine, 4,6-dichloropyrimidine methyl sulphide, 2-methylthio-4,6-dichloropyrimidine, pubchem21477
PubChem CID 80531
IUPAC Name 4,6-dichloro-2-methylsulfanylpyrimidine
SMILES CSC1=NC(=CC(=N1)Cl)Cl

Specifications

Melting Point 42°C
Color White-Yellow
Boiling Point 136°C
Quantity 5 g
UN Number 1759
Formula Weight 195.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dichloro-2-(methylthio)pyrimidine
TSCA No
Recommended Storage Refrigerator