accessibility menu, dialog, popup

UserName

H3CSNClClN

CAS RN 6299-25-8

IUPAC Name: 4,6-dichloro-2-(methylsulfanyl)pyrimidine
Synonyms: 4,6-dichloro-2-(methylsulfanyl)pyrimidine
Molecular Weight (g/mol): 195.06
Molecular Formula: C5H4Cl2N2S
InChi Key: FCMLONIWOAGZJX-UHFFFAOYSA-N
SMILES: CSC1=NC(Cl)=CC(Cl)=N1
Boiling Point 
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)