missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-2-(methylthio)pyrimidine, 98%, Thermo Scientific™

Chemical Identifiers

CAS 6299-25-8
Molecular Formula C5H4Cl2N2S
Molecular Weight (g/mol) 195.07
MDL Number MFCD00006086
InChI Key FCMLONIWOAGZJX-UHFFFAOYSA-N
Synonym 4,6-dichloro-2-methylthio pyrimidine, 4,6-dichloro-2-methylmercaptopyrimidine, 4,6-dichloro-2-methylsulfanyl-pyrimidine, 4,6-dichloro-2-methylthiopyrimidine, pyrimidine, 4,6-dichloro-2-methylthio, 4,6-dichloro-2-methylsulfanyl pyrimidine, 4,6-dichloropyrimidine methyl sulphide, 2-methylthio-4,6-dichloropyrimidine, pubchem21477
PubChem CID 80531
IUPAC Name 4,6-dichloro-2-methylsulfanylpyrimidine
SMILES CSC1=NC(=CC(=N1)Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC163030050
SDS View Documents
Thermo Scientific Chemicals
163030050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6299-25-8
195.07
FCMLONIWOAGZJX-UHFFFAOYSA-N
80531
CSC1=NC(=CC(=N1)Cl)Cl
C5H4Cl2N2S
MFCD00006086
4,6-dichloro-2-methylthio pyrimidine, 4,6-dichloro-2-methylmercaptopyrimidine, 4,6-dichloro-2-methylsulfanyl-pyrimidine, 4,6-dichloro-2-methylthiopyrimidine, pyrimidine, 4,6-dichloro-2-methylthio, 4,6-dichloro-2-methylsulfanyl pyrimidine, 4,6-dichloropyrimidine methyl sulphide, 2-methylthio-4,6-dichloropyrimidine, pubchem21477
4,6-dichloro-2-methylsulfanylpyrimidine

Specifications

6299-25-8
100.0
>112°C
97.5% min. (GC)
C5H4Cl2N2S
5g
4,6-dichloro-2-methylthio pyrimidine, 4,6-dichloro-2-methylmercaptopyrimidine, 4,6-dichloro-2-methylsulfanyl-pyrimidine, 4,6-dichloro-2-methylthiopyrimidine, pyrimidine, 4,6-dichloro-2-methylthio, 4,6-dichloro-2-methylsulfanyl pyrimidine, 4,6-dichloropyrimidine methyl sulphide, 2-methylthio-4,6-dichloropyrimidine, pubchem21477
CSC1=NC(=CC(=N1)Cl)Cl
195.07
195.07
4, 6-Dichloro-2-(methylthio)pyrimidine
97.5
135.0°C to 136.0°C (14.0mmHg)
Authentic
Glass bottle
MFCD00006086
23, 371
FCMLONIWOAGZJX-UHFFFAOYSA-N
4,6-dichloro-2-methylsulfanylpyrimidine
80531
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 228-577-6

SDS
Documents

Documents

RUO – Research Use Only