missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-2-(methylthio)pyrimidine, 98%

Catalog No. AC163030050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163030050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC163030050 Supplier Thermo Scientific Chemicals Supplier No. 163030050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6299-25-8 | C5H4Cl2N2S | 195.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6299-25-8
Molecular Formula C5H4Cl2N2S
Molecular Weight (g/mol) 195.07
MDL Number MFCD00006086
InChI Key FCMLONIWOAGZJX-UHFFFAOYSA-N
Synonym 4,6-dichloro-2-methylthio pyrimidine, 4,6-dichloro-2-methylmercaptopyrimidine, 4,6-dichloro-2-methylsulfanyl-pyrimidine, 4,6-dichloro-2-methylthiopyrimidine, pyrimidine, 4,6-dichloro-2-methylthio, 4,6-dichloro-2-methylsulfanyl pyrimidine, 4,6-dichloropyrimidine methyl sulphide, 2-methylthio-4,6-dichloropyrimidine, pubchem21477
PubChem CID 80531
IUPAC Name 4,6-dichloro-2-methylsulfanylpyrimidine
SMILES CSC1=NC(=CC(=N1)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 38.0°C to 42.0°C
Color Orange or White to Yellow
Boiling Point 135.0°C to 136.0°C (14.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 23, 371
Formula Weight 195.07
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 4, 6-Dichloro-2-(methylthio)pyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 228-577-6

RUO – Research Use Only