missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-2-(methylthio)pyrimidine, 98%, Thermo Scientific Chemicals

$139.29 - $139.29

Chemical Identifiers

CAS 6299-25-8
Molecular Formula C5H4Cl2N2S
Molecular Weight (g/mol) 195.061
MDL Number MFCD00006086
InChI Key FCMLONIWOAGZJX-UHFFFAOYSA-N
Synonym 4,6-dichloro-2-methylthio pyrimidine, 4,6-dichloro-2-methylmercaptopyrimidine, 4,6-dichloro-2-methylsulfanyl-pyrimidine, 4,6-dichloro-2-methylthiopyrimidine, pyrimidine, 4,6-dichloro-2-methylthio, 4,6-dichloro-2-methylsulfanyl pyrimidine, 4,6-dichloropyrimidine methyl sulphide, 2-methylthio-4,6-dichloropyrimidine, pubchem21477
PubChem CID 80531
IUPAC Name 4,6-dichloro-2-methylsulfanylpyrimidine
SMILES CSC1=NC(=CC(=N1)Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2141606
View Documents
Thermo Scientific Chemicals
B2141606
5 g
Each for $139.29
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6299-25-8
195.061
FCMLONIWOAGZJX-UHFFFAOYSA-N
80531
CSC1=NC(=CC(=N1)Cl)Cl
C5H4Cl2N2S
MFCD00006086
4,6-dichloro-2-methylthio pyrimidine, 4,6-dichloro-2-methylmercaptopyrimidine, 4,6-dichloro-2-methylsulfanyl-pyrimidine, 4,6-dichloro-2-methylthiopyrimidine, pyrimidine, 4,6-dichloro-2-methylthio, 4,6-dichloro-2-methylsulfanyl pyrimidine, 4,6-dichloropyrimidine methyl sulphide, 2-methylthio-4,6-dichloropyrimidine, pubchem21477
4,6-dichloro-2-methylsulfanylpyrimidine

Specifications

6299-25-8
135°C to 136°C (14 mmHg)
Irritating
MFCD00006086
UN1759
4,6-dichloro-2-methylthio pyrimidine, 4,6-dichloro-2-methylmercaptopyrimidine, 4,6-dichloro-2-methylsulfanyl-pyrimidine, 4,6-dichloro-2-methylthiopyrimidine, pyrimidine, 4,6-dichloro-2-methylthio, 4,6-dichloro-2-methylsulfanyl pyrimidine, 4,6-dichloropyrimidine methyl sulphide, 2-methylthio-4,6-dichloropyrimidine, pubchem21477
CSC1=NC(=CC(=N1)Cl)Cl
195.061
195.07
4,6-Dichloro-2-(methylthio)pyrimidine
38°C to 42°C
>110°C (230°F)
C5H4Cl2N2S
5 g
127664
FCMLONIWOAGZJX-UHFFFAOYSA-N
4,6-dichloro-2-methylsulfanylpyrimidine
80531
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H500

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

EINECSNumber : 228-577-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only