missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Dichloro-2-(methylsulfonyl)pyrimidine 98.0+%, TCI America™
Supplier: TCI America D41355G
Specifications
| 4,6-Dichloro-2-(methylsulfonyl)pyrimidine | |
| 120°C | |
| MFCD00672147 | |
| 4,6-Dichloro-2-(methanesulfonyl)pyrimidine | |
| CS(=O)(=O)C1=NC(=CC(=N1)Cl)Cl | |
| 227.059 | |
| 227.06 | |
| Crystalline Powder |
| 4489-34-3 | |
| C5H4Cl2N2O2S | |
| 5 g | |
| DROUVIKCNOHKBA-UHFFFAOYSA-N | |
| 4,6-dichloro-2-methylsulfonylpyrimidine | |
| 239685 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 4489-34-3 | |
| 227.059 | |
| DROUVIKCNOHKBA-UHFFFAOYSA-N | |
| 239685 | |
| CS(=O)(=O)C1=NC(=CC(=N1)Cl)Cl |
| C5H4Cl2N2O2S | |
| MFCD00672147 | |
| 4,6-Dichloro-2-(methanesulfonyl)pyrimidine | |
| 4,6-dichloro-2-methylsulfonylpyrimidine |
Safety and Handling
TSCA : No