accessibility menu, dialog, popup

UserName

CAS RN 4489-34-3

CH3SOONClClN

CAS RN 4489-34-3

IUPAC Name: 4,6-dichloro-2-methanesulfonylpyrimidine
Synonyms: 4,6-dichloro-2-methanesulfonylpyrimidine
Molecular Weight (g/mol): 227.06
Molecular Formula: C5H4Cl2N2O2S
InChi Key: DROUVIKCNOHKBA-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=NC(Cl)=CC(Cl)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results