missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-2-methylpyrimidine 98.0+%, TCI America™

Catalog No. p-7127791
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35585G 5 g
D355825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35585G Supplier TCI America Supplier No. D35585G

Chemical Identifiers

CAS 1780-26-3
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.001
MDL Number MFCD00090472
InChI Key FIMUTBLUWQGTIJ-UHFFFAOYSA-N
Synonym 2-methyl-4,6-dichloropyrimidine, 4,6-dichloro-2-methyl-pyrimidine, 2-mdcp, pyrimidine, 4,6-dichloro-2-methyl, 4,6-dichloro-2-methylpyrimidiine, 4,6-dichlor-2-methylpyrimidin, 4,6-dichloro-2-methyl pyrimidine, pubchem7075, acmc-1bpg6, ksc490s3h
PubChem CID 234997
IUPAC Name 4,6-dichloro-2-methylpyrimidine
SMILES CC1=NC(=CC(=N1)Cl)Cl

Specifications

Melting Point 47°C
Color White-Yellow
Quantity 5 g
Formula Weight 163.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dichloro-2-methylpyrimidine