missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dibromopyrimidine 98.0+%, TCI America™

Catalog No. p-7128361
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46001G 1 g
D46005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46001G Supplier TCI America Supplier No. D46001G

Chemical Identifiers

CAS 36847-10-6
Molecular Formula C4H2Br2N2
Molecular Weight (g/mol) 237.88
MDL Number MFCD00233946
InChI Key CEPVZSXIONCAJV-UHFFFAOYSA-N
PubChem CID 20085404
IUPAC Name 4,6-dibromopyrimidine
SMILES BrC1=CC(Br)=NC=N1

Specifications

Melting Point 52°C
Color White-Yellow
Quantity 1 g
Synonym 4,6-dibromo-pyrimidine, pyrimidine, 4,6-dibromo, 4,6-dibromopyrimydine, pyrimidine,4,6-dibromo, acmc-1adp1
Formula Weight 237.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dibromopyrimidine