accessibility menu, dialog, popup

UserName

CAS RN 36847-10-6

IUPAC Name: 4,6-dibromopyrimidine
Synonyms: 4,6-dibromopyrimidine
Molecular Weight (g/mol): 237.88
Molecular Formula: C4H2Br2N2
InChi Key: CEPVZSXIONCAJV-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=NC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results