missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diaminoresorcinol dihydrochloride, 98%

Catalog No. p-2690544
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC333670050 5 g Glass bottle
AC333670250 25 g Glass bottle
AC333671000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC333670050 Supplier Thermo Scientific Chemicals Supplier No. 333670050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16523-31-2 | C6H8N2O2·2HCl | 213.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16523-31-2
Molecular Formula C6H8N2O2·2HCl
Molecular Weight (g/mol) 213.06
MDL Number MFCD00143239
InChI Key KUMOYHHELWKOCB-UHFFFAOYSA-N
Synonym 4,6-diaminoresorcinol dihydrochloride, 4,6-diaminobenzene-1,3-diol dihydrochloride, 4,6-diaminoresorcinol 2hcl, 1,3-benzenediol, 4,6-diamino-, dihydrochloride, 4,6-diamino resorcinol dihydrochloride, 4,6-diamino-benzene-1,3-diol 2hcl salt, 1,3-benzenediol, 4,6-diamino-, hydrochloride 1:2, 4,6-bis azanyl benzene-1,3-diol dihydrochloride, pubchem20873, acmc-1c78d
PubChem CID 2733648
IUPAC Name 4,6-diaminobenzene-1,3-diol;dihydrochloride
SMILES C1=C(C(=CC(=C1N)O)O)N.Cl.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown-Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 13,I,317
Formula Weight 213.06
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4, 6-Diaminoresorcinol dihydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only