missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diamino-2-(methylthio)pyrimidine, 97%, Thermo Scientific Chemicals

Catalog No. p-7044707
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0788814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0788814 Supplier Thermo Scientific Chemicals Supplier No. L0788814
Only null left

4,6-Diamino-2-(methylthio)pyrimidine can reacts with 3-Ethoxy-acrylic acid ethyl ester to get 4-Amino-2-methylsulfanyl-8H-pyrido[2,3-d]pyrimidin-7-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Diamino-2-(methylthio)pyrimidine can reacts with 3-Ethoxy-acrylic acid ethyl ester to get 4-Amino-2-methylsulfanyl-8H-pyrido[2,3-d]pyrimidin-7-one.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 1005-39-6
Molecular Formula C5H8N4S
Molecular Weight (g/mol) 156.207
MDL Number MFCD00023241
InChI Key AHAIUNAIAHSWPG-UHFFFAOYSA-N
Synonym 4,6-diamino-2-methylmercaptopyrimidine, 2-methylthio pyrimidine-4,6-diamine, 2-methylthiopyrimidine-4,6-diamine, 4,6-diamino-2-methylthiopyrimidine, 4,6-pyrimidinediamine, 2-methylthio, 4,6-diamino-2-methylthio pyrimidine, 2-methylsulfanyl pyrimidine-4,6-diamine, 2-methylthio-4,6-diaminopyrimidine, 4,6-diamino-2-methyl-mercaptopyrimidine, pyrimidine, 4,6-diamino-2-methylthio
PubChem CID 70497
IUPAC Name 2-methylsulfanylpyrimidine-4,6-diamine
SMILES CSC1=NC(=CC(=N1)N)N

Specifications

Melting Point 187°C to 191°C
Quantity 25 g
Beilstein 127235
Formula Weight 156.21
Percent Purity 97%
Chemical Name or Material 4,6-Diamino-2-(methylthio)pyrimidine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 213-735-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only