missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diacetylresorcinol 98.0+%, TCI America™

Catalog No. D51025G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D51025G Supplier TCI America Supplier No. D51025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2161-85-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00017655
InChI Key GEYCQLIOGQPPFM-UHFFFAOYSA-N
Synonym 4,6-diacetylresorcinol, resodiacetophenone, 1,1'-4,6-dihydroxy-1,3-phenylene bisethanone, ethanone, 1,1'-4,6-dihydroxy-1,3-phenylene bis, 1-5-acetyl-2,4-dihydroxyphenyl ethanone, 1,1'-4,6-dihydroxy-1,3-phenylene diethanone, 1,3-diacetyl-4,6-dihydroxybenzene, ethanone,1,1'-4,6-dihydroxy-1,3-phenylene bis, 1-5-acetyl-2,4-dihydroxyphenyl ethan-1-one, 1,1'-4,6-dihydroxy-1,3-phenylene-bis-ethanone
PubChem CID 75097
IUPAC Name 1-(5-acetyl-2,4-dihydroxyphenyl)ethan-1-one
SMILES CC(=O)C1=CC(C(C)=O)=C(O)C=C1O

Specifications

Melting Point 184°C
Quantity 5 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Diacetylresorcinol