accessibility menu, dialog, popup

UserName

H3COCH3OOHHO

CAS RN 2161-85-5

IUPAC Name: 1-(5-acetyl-2,4-dihydroxyphenyl)ethan-1-one
Synonyms: 1-(5-acetyl-2,4-dihydroxyphenyl)ethanone
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: GEYCQLIOGQPPFM-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(C(C)=O)=C(O)C=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results