missing translation for 'onlineSavingsMsg'
Learn More

4,6-Bis(difluoromethoxy)-2-(methylthio)pyrimidine 98.0+%, TCI America™

Catalog No. B275525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B275525G 25 g
B27555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B275525G Supplier TCI America Supplier No. B275525G

Chemical Identifiers

CAS 100478-25-9
Molecular Formula C7H6F4N2O2S
Molecular Weight (g/mol) 258.191
InChI Key HURDQHYOXAOOJJ-UHFFFAOYSA-N
PubChem CID 11043427
IUPAC Name 4,6-bis(difluoromethoxy)-2-methylsulfanylpyrimidine
SMILES CSC1=NC(=CC(=N1)OC(F)F)OC(F)F

Specifications

Melting Point 48°C
Color White-Yellow
Quantity 25 g
Formula Weight 258.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Bis(difluoromethoxy)-2-(methylthio)pyrimidine
EINECSNumber (5)-6194
TSCA No