missing translation for 'onlineSavingsMsg'
Learn More

4,6,8-Trimethylquinoline 98.0+%, TCI America™

Catalog No. T14091G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T14091G Supplier TCI America Supplier No. T14091G
Only null left

Chemical Identifiers

CAS 88565-88-2
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.243
MDL Number MFCD00191505
InChI Key HNCYZWOTJQPKTQ-UHFFFAOYSA-N
PubChem CID 44630515
IUPAC Name 4,6,8-trimethylquinoline
SMILES CC1=C2C=C(C=C(C2=NC=C1)C)C

Specifications

Melting Point 57°C
Quantity 1 g
Formula Weight 171.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,6,8-Trimethylquinoline