accessibility menu, dialog, popup

UserName

CAS RN 88565-88-2

CH3NCH3CH3

CAS RN 88565-88-2

IUPAC Name: 4,6,8-trimethylquinoline
Synonyms: 4,6,8-trimethylquinoline
Molecular Weight (g/mol): 171.24
Molecular Formula: C12H13N
InChi Key: HNCYZWOTJQPKTQ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C2N=CC=C(C)C2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results