missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Ethylenedithio-1,3-dithiole-2-thione 98.0+%, TCI America™
Supplier: TCI America E042925G
Specifications
| 4,5-Ethylenedithio-1,3-dithiole-2-thione | |
| 124°C | |
| MFCD00137882 | |
| LFPIFRGVCYRUEO-UHFFFAOYSA-N | |
| 5,6-dihydro-[1,3]dithiolo[4,5-b][1,4]dithiine-2-thione | |
| 360963 | |
| ≥98.0% (GC) |
| 59089-89-3 | |
| C5H4S5 | |
| 25 g | |
| C1CSC2=C(S1)SC(=S)S2 | |
| 224.387 | |
| 224.39 | |
| Crystalline Powder |
Chemical Identifiers
| 59089-89-3 | |
| 224.387 | |
| LFPIFRGVCYRUEO-UHFFFAOYSA-N | |
| 5,6-dihydro-[1,3]dithiolo[4,5-b][1,4]dithiine-2-thione |
| C5H4S5 | |
| MFCD00137882 | |
| 360963 | |
| C1CSC2=C(S1)SC(=S)S2 |
Safety and Handling
TSCA : No