missing translation for 'onlineSavingsMsg'
Learn More

4,5-Ethylenedithio-1,3-dithiole-2-thione 98.0+%, TCI America™

Catalog No. p-7128992
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04291G 1 g
E04295G 5 g
E042925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E04291G Supplier TCI America Supplier No. E04291G
Only null left

Chemical Identifiers

CAS 59089-89-3
Molecular Formula C5H4S5
Molecular Weight (g/mol) 224.387
MDL Number MFCD00137882
InChI Key LFPIFRGVCYRUEO-UHFFFAOYSA-N
PubChem CID 360963
IUPAC Name 5,6-dihydro-[1,3]dithiolo[4,5-b][1,4]dithiine-2-thione
SMILES C1CSC2=C(S1)SC(=S)S2

Specifications

Melting Point 124°C
Quantity 1 g
Formula Weight 224.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Ethylenedithio-1,3-dithiole-2-thione