missing translation for 'onlineSavingsMsg'
Learn More

4,5-Ethylenedithio-1,3-dithiol-2-one 98.0+%, TCI America™

Catalog No. p-7129010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04601G 1 g
E04605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04601G Supplier TCI America Supplier No. E04601G

Chemical Identifiers

CAS 74962-29-1
Molecular Formula C5H4OS4
Molecular Weight (g/mol) 208.33
MDL Number MFCD00137883
InChI Key NMNDMYIFMUGDMA-UHFFFAOYSA-N
PubChem CID 610302
IUPAC Name 2H,5H,6H-[1,3]dithiolo[4,5-b][1,4]dithiin-2-one
SMILES O=C1SC2=C(S1)SCCS2

Specifications

Melting Point 129°C
Quantity 1 g
Formula Weight 208.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Ethylenedithio-1,3-dithiol-2-one