accessibility menu, dialog, popup

UserName

CAS RN 74962-29-1

IUPAC Name: 2H,5H,6H-[1,3]dithiolo[4,5-b][1,4]dithiin-2-one
Synonyms: 5H,6H-[1,3]dithiolo[4,5-b][1,4]dithiin-2-one
Molecular Weight (g/mol): 208.33
Molecular Formula: C5H4OS4
InChi Key: NMNDMYIFMUGDMA-UHFFFAOYSA-N
SMILES: O=C1SC2=C(S1)SCCS2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results