missing translation for 'onlineSavingsMsg'
Learn More

4,5-Diphenyl-2-oxazolethiol 98.0+%, TCI America™

Catalog No. D251125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D251125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D251125G Supplier TCI America Supplier No. D251125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6670-13-9
Molecular Formula C15H11NOS
Molecular Weight (g/mol) 253.319
MDL Number MFCD00173660
InChI Key NYQSTRUSYDZNHC-UHFFFAOYSA-N
Synonym 4,5-diphenyl-2-oxazolethiol, 4,5-diphenyl-1,3-oxazole-2-thiol, diphenyl-1,3-oxazole-2-thiol, 4,5-diphenyl-2-mercaptooxazole, 4,5-diphenyl-1,3-oxazole-2 3h-thione, 4,5-diphenyloxazole-2 3h-thione, 4,5-diphenyl-4-oxazoline-2-thione, 4,5-diphenyl-2,3-dihydro-1,3-oxazole-2-thione, 4,5-diphenyloxazole-2-thiol, 4,5-diphenyl-3h-oxazole-2-thione
PubChem CID 853676
IUPAC Name 4,5-diphenyl-3H-1,3-oxazole-2-thione
SMILES C1=CC=C(C=C1)C2=C(OC(=S)N2)C3=CC=CC=C3

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 253.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Diphenyl-2-oxazolethiol