CAS RN 6670-13-9
IUPAC Name:
4,5-diphenyl-2,3-dihydro-1,3-oxazole-2-thione
Synonyms:
4,5-diphenyl-3H-1,3-oxazole-2-thione
Molecular Weight (g/mol):
253.32
Molecular Formula:
C15H11NOS
InChi Key:
NYQSTRUSYDZNHC-UHFFFAOYSA-N
SMILES:
S=C1NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
4,5-Diphenyl-2-oxazolethiol 98.0+%, TCI America™
CAS: 6670-13-9 Molecular Formula: C15H11NOS Molecular Weight (g/mol): 253.319 MDL Number: MFCD00173660 InChI Key: NYQSTRUSYDZNHC-UHFFFAOYSA-N Synonym: 4,5-diphenyl-2-oxazolethiol,4,5-diphenyl-1,3-oxazole-2-thiol,diphenyl-1,3-oxazole-2-thiol,4,5-diphenyl-2-mercaptooxazole,4,5-diphenyl-1,3-oxazole-2 3h-thione,4,5-diphenyloxazole-2 3h-thione,4,5-diphenyl-4-oxazoline-2-thione,4,5-diphenyl-2,3-dihydro-1,3-oxazole-2-thione,4,5-diphenyloxazole-2-thiol,4,5-diphenyl-3h-oxazole-2-thione PubChem CID: 853676 IUPAC Name: 4,5-diphenyl-3H-1,3-oxazole-2-thione SMILES: C1=CC=C(C=C1)C2=C(OC(=S)N2)C3=CC=CC=C3