missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dimethylthiazole 98.0+%, TCI America™
Supplier: TCI America D214225G
Specifications
| 4,5-Dimethylthiazole | |
| Yellow | |
| C5H7NS | |
| 25 g | |
| 4,5-dimethylthiazole, thiazole, 4,5-dimethyl, 4,5-dimethyl thiazole, 4,5-dimethyl-thiazole, unii-u3rp5i088g, fema no. 3274, pubchem9938, acmc-209ijb, dimethylthiazole, 4,5, 4-27-00-00986 beilstein handbook reference | |
| CC1=C(SC=N1)C | |
| 113.178 | |
| 113.18 | |
| Liquid |
| 3581-91-7 | |
| 159°C | |
| MFCD00005336 | |
| 1993 | |
| UWSONZCNXUSTKW-UHFFFAOYSA-N | |
| 4,5-dimethyl-1,3-thiazole | |
| 62510 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3581-91-7 | |
| 113.178 | |
| UWSONZCNXUSTKW-UHFFFAOYSA-N | |
| 62510 | |
| CC1=C(SC=N1)C |
| C5H7NS | |
| MFCD00005336 | |
| 4,5-dimethylthiazole, thiazole, 4,5-dimethyl, 4,5-dimethyl thiazole, 4,5-dimethyl-thiazole, unii-u3rp5i088g, fema no. 3274, pubchem9938, acmc-209ijb, dimethylthiazole, 4,5, 4-27-00-00986 beilstein handbook reference | |
| 4,5-dimethyl-1,3-thiazole |
Safety and Handling
RTECSNumber : XJ4380000
TSCA : No