missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethylthiazole 98.0+%, TCI America™

Catalog No. p-7127131
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
D21425G 5 g
D214225G 25 g
2 options

Catalog No. D21425G

Supplier: TCI America D21425G

Only null left

Chemical Identifiers

CAS 3581-91-7
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.178
MDL Number MFCD00005336
InChI Key UWSONZCNXUSTKW-UHFFFAOYSA-N
Synonym 4,5-dimethylthiazole, thiazole, 4,5-dimethyl, 4,5-dimethyl thiazole, 4,5-dimethyl-thiazole, unii-u3rp5i088g, fema no. 3274, pubchem9938, acmc-209ijb, dimethylthiazole, 4,5, 4-27-00-00986 beilstein handbook reference
PubChem CID 62510
IUPAC Name 4,5-dimethyl-1,3-thiazole
SMILES CC1=C(SC=N1)C

Specifications

Color Yellow
Boiling Point 159°C
Quantity 5 g
UN Number 1993
Formula Weight 113.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,5-Dimethylthiazole
RTECSNumber XJ4380000
TSCA No