missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethyl-o-phenylenediamine, 98%

Catalog No. p-4229189
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC116610050 5 g Glass bottle
AC116610250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC116610050 Supplier Thermo Scientific Chemicals Supplier No. 116610050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3171-45-7 | C8H12N2 | 136.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3171-45-7
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD00007729
InChI Key XSZYBMMYQCYIPC-UHFFFAOYSA-N
Synonym 4,5-dimethyl-1,2-phenylenediamine, 4,5-diamino-o-xylene, 4,5-dimethyl-o-phenylenediamine, 1,2-benzenediamine, 4,5-dimethyl, 1,2-diamino-4,5-dimethylbenzene, 4,5-dimethyl-1,2-benzenediamine, 4,5-dimethyl-benzene-1,2-diamine, 4,5-dimethyl-ortho-phenylenediamine, 4,5-dimethylphenylene-1,2-diamine, 4,5-dimethyl-o-phenylenediamine monohydrate
PubChem CID 76635
IUPAC Name 4,5-dimethylbenzene-1,2-diamine
SMILES CC1=CC(N)=C(N)C=C1C

Specifications

Melting Point 127.2°C to 128.3°C
Color Brown to White
CAS Min % 97.5
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Mass, Crystals or Chunks
Assay Percent Range 97.5% min. (GC)
Linear Formula (CH3)2C6H2(NH2)2
Beilstein 13,179
Solubility Information Solubility in water: slightly soluble. Other solubilities: good soluble in acidified water,soluble in alcohols and polar solvents
Formula Weight 136.19
Percent Purity 98%
Chemical Name or Material 4, 5-Dimethyl-o-phenylenediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 221-635-1

RUO – Research Use Only