missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethyl-2-nitroaniline, 97%

Catalog No. AAA1555622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1555622 100 g
AAA1555606 5 g
AAA1555614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1555622 Supplier Thermo Scientific Chemicals Supplier No. A1555622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6972-71-0 | C8H10N2O2 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6972-71-0
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00007811
InChI Key PINGKGKKUSYUAW-UHFFFAOYSA-N
Synonym 6-nitro-3,4-xylidine, benzenamine, 4,5-dimethyl-2-nitro, 3,4-xylidine, 6-nitro, 2-nitro-4,5-dimethylaniline, 4,5-dimethyl-2-nitrophenylamine, 2-nitro-4,5-xylidine, 3,4-dimethyl-6-nitroaniline, acmc-20amrx, 3, 6-nitro, 4-amino-5-nitro-o-xylene
PubChem CID 81445
IUPAC Name 4,5-dimethyl-2-nitroaniline
SMILES CC1=CC(=C(C=C1C)[N+](=O)[O-])N

Specifications

Melting Point 138°C to 143°C
Quantity 100 g
UN Number UN2811
Beilstein 2209637
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 4,5-Dimethyl-2-nitroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 230-211-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only