missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dimethyl-1,2-phenylenediamine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 127°C |
| Quantity | 5 g |
| Physical Form | Crystal-Powder at 20°C |
| Formula Weight | 136.20 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Not Available |
| Chemical Name or Material | 4,5-Dimethyl-1,2-phenylenediamine |
Safety and Handling
| TSCA | Yes |