missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate 90.0+%, TCI America™

Catalog No. D38831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38831G 1 g
D38835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38831G Supplier TCI America Supplier No. D38831G

Chemical Identifiers

CAS 866252-52-0
Molecular Formula C12H17F3O5SSi
Molecular Weight (g/mol) 358.40
MDL Number MFCD15072159
InChI Key DLUVZOFWZNLGTJ-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid 4,5-Dimethoxy-2-(trimethylsilyl)phenyl Ester, 4,5-Dimethoxy-2-(trimethylsilyl)phenyl Triflate
PubChem CID 16120028
IUPAC Name 4,5-dimethoxy-2-(trimethylsilyl)phenyl trifluoromethanesulfonate
SMILES COC1=CC(OS(=O)(=O)C(F)(F)F)=C(C=C1OC)[Si](C)(C)C

Specifications

Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 358.40
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,5-Dimethoxy-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate
TSCA No
Recommended Storage Freezer