missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzyl chloroformate, 97%

Catalog No. AAL1416703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1416703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1416703 Supplier Thermo Scientific Chemicals Supplier No. L1416703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42855-00-5 | C10H10ClNO6 | 275.641 g/mol

4,5-Dimethoxy-2-nitrobenzyl chloroformate is used as a reagent for protection of amines, for use in N-protection of bases in nucleotide synthesis, photochemical protecting groups in organic synthesis. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dimethoxy-2-nitrobenzyl chloroformate is used as a reagent for protection of amines, for use in N-protection of bases in nucleotide synthesis, photochemical protecting groups in organic synthesis. It is used as pharmaceutical intermediate.

Solubility
Soluble in methanol, acetone. Reacts with water.

Notes
Moisture and light sensitive. Store away from oxidizing agents, light, water/ moisture, heat, bases.

Chemical Identifiers

CAS 42855-00-5
Molecular Formula C10H10ClNO6
Molecular Weight (g/mol) 275.641
MDL Number MFCD00143507
InChI Key RWWPKIOWBQFXEE-UHFFFAOYSA-N
Synonym 4,5-dimethoxy-2-nitrobenzyl carbonochloridate, nvoc-cl, 4,5-dimethoxy-2-nitrobenzyl chloroformate, 6-nitroveratryl chloroformate, nitroveratryl oxy chlorocarbamate, 6-nitroveratryloxycarbonyl chloride, carbonochloridic acid, 4,5-dimethoxy-2-nitrophenyl methyl ester, 4,5-dimethoxy-2-nitrophenyl methyl chloroformate, 4,5-dimethoxy-2-nitrophenyl methyl carbonochloridate, nvoc chloride
PubChem CID 3084878
IUPAC Name (4,5-dimethoxy-2-nitrophenyl)methyl carbonochloridate
SMILES COC1=C(C=C(C(=C1)COC(=O)Cl)[N+](=O)[O-])OC

Specifications

Melting Point ∽120°C (decomposition)
Quantity 1 g
UN Number UN3261
Beilstein 2389168
Sensitivity Light and moisture sensitive
Solubility Information Soluble in methanol,acetone. Reacts with water.
Formula Weight 275.64
Percent Purity 97%
Chemical Name or Material 4,5-Dimethoxy-2-nitrobenzyl chloroformate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only